Learn More
Waters Corp Waters Column, C18, 130 Å, 3.5 μm
Supplier: Waters Corp 186003567
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
- Chemistry: C18
- Separation mode: Reversed phase
- Particle substrate: Hybrid
- pH range: 1 to 12
- Maximum pressure: 6000 psi (415 Bar)
- Particle size: 3.5 μm
- Pore size: 130 Å
- Inner diameter: 4.6 mm
- Length: 50 mm
- System: HPLC
- Application: Peptide
- Carbon load: 17%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.