Learn More
Waters Corp WATERS CORPORATION Column, C18, 130Å 5 μm, 1 mm X 100 mm
Supplier: Waters Corp 186003572
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
- C18 chemistry for reversed phase separation.
- Hybrid, spherical particle substrate with 5 μm particle size.
- Dimensions: 1 mm inner diameter and 100 mm length.
- Maximum pressure of 6000 psi (415 Bar).
- Molecular weight range of 1000 to 15000.
- Endcapped with low silanol activity.
- Suitable for HPLC systems.
- USP Classification L1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.