Learn More
Waters Corp XBridge Peptide BEH C18 Prep Guard Cartridge, 130Å, 10 µm, 10 mm X 10 mm, 1/pk
Supplier:  Waters Corp 186004465
Encompass_Preferred
The XBridge BEH130 PREP C18, 10 µm, 10 x 10 mm Guard Column was develeped and designed to extend the life the BEH130, C18, 10um, 10mm ID preparative column. The BEH130 PREP C18, 10um packing material used in the Guard Column must also pass the peptide digest, QC test criteria established for the prep column.
Catalog No. 50-466-006