Learn More
Waters Corp Waters Column, C18, 130Å, 10 μm, 4.6 mm X 50 mm
Supplier:  Waters Corp 186003648
Encompass_Preferred
XBridge BEH130 C18 Peptide Separation Technology is optimized and QC tested for peptide separations. Small-pore (130Å) trifunctionally bonded BEH particles offer a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Optimized for peptide separations, QC tested.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Wide usable pH range (1 to 12 pH).
  • Superior low pH stability; ultra-low column bleed.
  • Reliable batch-to-batch separation of synthetic peptides/protein digests.
  • Stable structure and bonding at high temperatures.
  • Good peak form, high efficiency, and stability.
  • cGMP, ISO 9002 manufacturing with ultra-pure reagents.
  • Batch certified with peptide separation for repeatable performance.
Catalog No. 50-781-920