Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Guard Column, C18, 5 μm, 130 Å, 10 mm x 10 mm

Supplier:  Waters Corp 186004469

Encompass_Preferred

These XBridge BEH130 C18 Peptide Separation Technology columns are engineered for high-performance peptide separations. Optimized and QC tested, they feature a small-pore (130Å) trifunctionally bonded BEH particle, delivering the widest usable pH range, superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.

  • Optimized and QC tested for peptide separations
  • Trifunctionally bonded BEH particle
  • Wide pH range (1 to 12) and superior low pH stability
  • Ultra-low column bleed
  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle size: 5 μm
  • Pore size: 130 Å
  • Maximum pressure: 6000 psi (415 Bar)
  • Molecular weight range: 1000 to 30000
  • Format: Guard column
  • System: HPLC

Catalog No. 50-782-441


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.