Learn More
Waters Corp XBridge Peptide BEH C18 OBD Prep Column, 130Å, 10 µm, 30 mm X 250 mm, 1/pk
Supplier:  Waters Corp 186003662
Encompass_Preferred
The XBridge BEH130 C18 Peptide Separation Technology preparative columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis. The Optimum Bed Denisity (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
Catalog No. 50-781-934
Add to Cart
Request a Quote