Learn More
Waters Corp Waters Column, C18, 300 Å, 3.5 μm, 2.1 mm x 50 mm
Supplier: Waters Corp 186003607
These columns are optimized and quality control (QC) tested for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle that provides the widest usable pH range, superior low pH stability, and ultra-low column bleed for analyzing samples in proteomics, protein characterization, and peptide synthesis.
- C18 chemistry.
- Reversed phase separation mode.
- Hybrid particle substrate.
- pH range of 1 to 12.
- Maximum pressure of 6000 psi (415 bar).
- Molecular weight range from 1000 to 15000.
- Particle size of 3.5 μm.
- Pore size of 300 Å.
- Inner diameter of 2.1 mm.
- Length of 50 mm.
- QC tested for peptide applications.
- Superior low pH stability.
- Ultra-low column bleed.
- Provides excellent peak shape, high efficiency, and stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.