Learn More
Waters Corp XBridge Peptide BEH C18 Column, 300Å, 5 µm, 1 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003617
Encompass_Preferred
The XBridge BEH300 C18 Peptide Separation Technology columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Catalog No. 50-781-893