Learn More
Waters Corp XBridge Column, C18, 5 μm, 300 Å, 4.6 mm × 50 mm
Supplier:  Waters Corp 186003622
Encompass_Preferred
XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers a wide usable pH range, superior low pH stability, and ultra-low column bleed. This high-performance reversed-phase column ensures precision and efficiency.
  • Optimized for peptide separations.
  • Wide-pore (300Å) trifunctionally bonded particle.
  • Wide pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Good peak shape and resolution for peptide/protein analysis.
  • 5 μm particles for high separation efficiency.
  • 4.6 mm inner diameter allows high flow rates.
  • 50 mm length provides long column life.
  • End-capped to minimize MS bleed, enhancing sensitivity.
Catalog No. 50-781-898