Learn More
Waters Corp XBridge Protein BEH C4 Column, 300 Å, 3.5 μm
Supplier: Waters Corp 186004500
This column is QC tested and optimized to separate proteins based on size, hydrophobicity, and isoelectric point. Its C4 ligand is less retentive compared to traditional C18 phases, which helps minimize protein carryover, increase protein recovery, and improve peak capacity.
- Chemistry: C4
- Separation mode: Reversed phase
- Particle substrate: Hybrid
- pH range: 2 to 10 pH
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped: No
- Silanol activity: Low
- Molecular weight range supported: 10,000 to 500,000
- Particle shape: Spherical
- Particle size: 3.5 μm
- Pore size: 300 Å
- QC tested for protein applications
- Format: Column
- Surface area: 90
- System: HPLC
- Particle technology: BEH
- USP classification: L26
- Inner diameter: 2.1 mm
- Length: 150 mm
- Carbon load: 8 %
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.