Learn More
Waters Corp Waters XBridge C4 Column, Protein, 300 Å, 3.5 μm
Supplier:  Waters Corp 186004498
Encompass_Preferred
This column is quality control (QC) tested and optimized for separating proteins based on size, hydrophobicity, and isoelectric point. Its C4 ligand offers less retention compared to traditional C18 phases, which helps minimize protein carryover, increase protein recovery, and improve peak capacity.
  • Designed for high-resolution separations of proteins and peptides with improved recovery and minimal sample loss.
  • 300Å pore size is optimized for the analysis of large biomolecules.
  • C4 bonded phase is suitable for the analysis of hydrophobic proteins and peptides.
  • Compatible with a wide pH range and diverse mobile phases, including organic modifiers.
  • Accommodates high molecular weights without compromising separation efficiency.
  • Recommended operating pH range is 1 to 12.
Catalog No. 50-782-446