Learn More
Waters Corp Waters XBridge Column, Peptide BEH C18, 300 Å, 10 μm
Supplier:  Waters Corp 186003664
Encompass_Preferred
These columns are specifically optimized and quality control (QC) tested for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle, which provides several benefits. These characteristics make the columns suitable for assaying samples in various applications such as proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Wide-pore (300Å) trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior stability at low pH.
  • Ultra-low column bleed.
Catalog No. 50-781-936