Learn More
Waters Corp WATERS CORPORATION XBridge Prep Column, C18, 300Å, 10 μm
Supplier: Waters Corp 186003677
The BEH300 C18 Peptide Separation Technology (PST) preparative columns are optimized for peptide separations. Featuring a wide-pore (300Å) trifunctionally bonded BEH particle, they offer a wide pH range, superior low pH stability, and ultra-low column bleed. This makes them suitable for proteomics, protein characterization, and peptide synthesis. The OBD hardware format ensures superior column lifetime.
- Chemistry: C18
- Separation mode: Reversed Phase
- Particle substrate: Hybrid
- pH range: 1 to 12
- Maximum pressure: 6000 psi (415 Bar)
- Particle size: 10 μm
- Pore size: 300 Å
- Format: OBD Prep Column
- Inner diameter: 30 mm
- Length: 250 mm
- Application: Peptide
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.