Learn More
Waters Corp Waters Prep Column, C18, 10 μm, 30 mm X 150 mm, 300 Å
Supplier: Waters Corp 186003676
The XBridge Peptide BEH C18 OBD Prep Column is designed for optimized peptide separations in proteomics, protein characterization, and peptide synthesis. It features a wide-pore (300Å) trifunctionally bonded BEH particle, offering a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed. Its OBD hardware ensures extended column lifetime, and it is QC tested with a peptide map for consistent performance.
- Chemistry: C18
- Separation mode: Reversed phase
- pH range: 1 to 12
- Maximum pressure: 6000 psi (415 Bar)
- Molecular weight range: 1000 to 15000
- Particle size: 10 μm
- Pore size: 300 Å
- Inner diameter: 30 mm
- Length: 150 mm
- System: HPLC
- USP classification: L1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.