Learn More
Waters Corp Waters XBridge Peptide BEH C18 OBD Prep Column
Supplier:  Waters Corp 186003672
Encompass_Preferred
The XBridge BEH300 C18 Peptide Separation Technology (PST) preparative columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis. The Optimum Bed Density (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 12
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 10 μm particle size
  • HPLC system compatibility
  • L1 USP classification
  • 19 mm inner diameter
  • 150 mm length
  • 12% carbon load
  • One unit per package
Catalog No. 50-781-944