Learn More
Waters Corp Waters XBridge BEH C18 Column, 130Å, 3.5 μm, 1 mm x 100 mm
Supplier:  Waters Corp 186003127
Encompass_Preferred
The XBridge BEH C18 sorbent is designed for HPLC method development, offering functionality across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand provides a universal column suitable for most HPLC separations, featuring a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Can be used over an entire range of mobile phase pH (1 to 12)
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations
  • Offers the widest usable pH range
  • Superior low pH stability
  • Ultra-low column bleed
  • C18 chemistry for reversed phase separation mode
  • Hybrid particle substrate with spherical particle shape
  • Maximum pressure of 6000 psi (415 bar)
  • 3.5 μm particle size
  • 130 Å pore size
  • 1 × 100 mm dimensions (inner diameter × length)
Catalog No. 50-781-563