Learn More
Waters Corp XBridge Column, C18, 3.5 μm, 4.6 mm × 75 mm, 130Å
Supplier:  Waters Corp 186003032
Encompass_Preferred
The BEH C18 sorbent serves as a workhorse for HPLC method development and can be utilized across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a universal column suitable for most HPLC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Widest usable pH range (1 to 12)
  • Superior low pH stability
  • Ultra-low column bleed
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations
Catalog No. 50-781-485