Learn More
Waters Corp Waters Corporation XBridge BEH C18 Column, 130Å, 3.5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186003034
Encompass_Preferred
The XBridge BEH C18 Column is a universal workhorse for HPLC method development, offering a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand provides superior low pH stability and ultra-low column bleed, maximizing productivity and enabling seamless transferability to UPLC for preparative scaling.
  • Increased lifetimes due to superior pH stability.
  • Thoroughly tested for reproducibility.
  • Precise control of silanol activity minimizes peak tailing, enhancing peak shape and capacity.
  • Suitable for small molecule separations in pharmaceuticals, environmental testing, and food safety analysis.
  • 130Å pore size optimizes interaction for improved separation efficiency.
  • 3.5 μm particle size offers excellent resolution and manageable back pressure.
  • Chemically stable up to pH 12 for broad mobile phase conditions.
Catalog No. 50-781-487