Learn More
Waters Corp Waters XBridge BEH C18 Column, 5 μm, 4.6 mm X 30 mm
Supplier:  Waters Corp 186003135
Encompass_Preferred
The XBridge BEH C18 sorbent serves as a workhorse for HPLC method development and can be utilized across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a universal column suitable for most HPLC separations.
  • Can be used over an entire range of mobile phase pH (1 to 12) as a workhorse for HPLC method development.
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations.
  • Offers the widest usable pH range.
  • Provides superior low pH stability.
  • Features ultra-low column bleed.
Catalog No. 50-781-571