Learn More
Waters Corp Waters XBridge Column Kit, BEH C18, 3.5 μm, 4.6 mm × 100 mm
Supplier:  Waters Corp 186003769
Encompass_Preferred
The XBridge BEH C18 sorbent is designed for HPLC method development and can be used across a mobile phase pH range of 1–12. Its trifunctionally bonded C18 ligand offers a universal column for most HPLC separations, providing the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Widest usable pH range (1–12)
  • Superior low pH stability
  • Ultra-low column bleed
  • Universal column for most HPLC separations
  • Trifunctionally bonded C18 ligand
  • Spherical particle shape
  • Low silanol activity
Catalog No. 50-782-024