Learn More
Waters Corp Waters XBridge Method Validation Kit, C18, 130Å, 3.5 μm
Supplier:  Waters Corp 186003766
Encompass_Preferred
This sorbent is a workhorse for HPLC method development, usable across a mobile phase pH range of 1-12. Its trifunctionally bonded C18 ligand offers a universal column for most HPLC separations, providing superior low pH stability and ultra-low column bleed. This kit defines conditions for demanding method development due to its wide pH range, offering optimum stability under extreme pH conditions.
  • Can be used over an entire range of mobile phase pH (1-12).
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations.
  • Offers superior low pH stability and ultra-low column bleed.
  • Allows quick definition of conditions for demanding method development.
  • Enables operation under any mobile phase, temperature, and pH conditions without compromising sorbent choice.
Catalog No. 50-782-021