Learn More
Waters Corp XBridge BEH C8 Column, 130Å, 3.5 µm, 1 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003179
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers you the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
Catalog No. 50-781-607