Learn More
Waters Corp Waters Column, C8, 3.5 μm, 2.1 mm ID, 150 mm
Supplier:  Waters Corp 186003049
Encompass_Preferred
The XBridge BEH C8 Column offers improved peak shape and chromatographic performance for demanding experiments. Its trifunctionally bonded sorbent ensures superior low pH stability, ultra-low column bleed, and a wide pH range (1 to 12), increasing laboratory output for LC/MS assays and quality control methods.
  • Offers less retention than C18 sorbents, enhancing speed and chromatographic performance.
  • Provides a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed due to trifunctionally bonded sorbent.
  • Delivers reliable findings and increases laboratory output for LC/MS assays and quality control.
  • Ensures assay robustness, improved pH stability, and a lengthy column lifetime.
  • Guarantees unmatched batch-to-batch and year-to-year reproducibility through careful testing.
Catalog No. 50-781-502