Learn More
Waters Corp Waters Column, C8, 3.5 μm, 130 Å, 3 mm x 100 mm
Supplier:  Waters Corp 186003051
Encompass_Preferred
The XBridge BEH C8 Column is designed for method development, offering less retention than C18 sorbents. Its trifunctionally bonded C8 sorbent provides a wide usable pH range (1-12), superior low pH stability, and ultra-low column bleed. This column enhances chromatographic performance and aids in efficient LC/MS and quality control method development, maximizing lab productivity.
  • Improves peak shape and chromatographic performance.
  • Features a wide usable pH range (1-12).
  • Provides superior low pH stability.
  • Ensures ultra-low column bleed.
  • Delivers consistently accurate results.
  • Maximizes lab productivity.
  • Offers enhanced column reliability and a long lifetime.
  • Achieves unmatched reproducibility.
  • Rigorously tested from raw material to final batch.
Catalog No. 50-781-504