Learn More
Waters Corp XBridge BEH C8 Column, 130Å, 3.5 μm, 4.6 mm × 30 mm
Supplier:  Waters Corp 186003184
Encompass_Preferred
The Waters XBridge BEH C8 Column incorporates Ethylene Bridged Hybrid (BEH) Technology, providing increased chemical and mechanical stability, and high pH stability for versatile chromatographic separations. It is designed for durable performance and long column lifetime under harsh operating conditions.
  • Versatile pH and temperature range for mobile phase.
  • Seamless method migration to UltraPerformance LC™ technology.
  • Unmatched column lifetime and stability across pH 1-12.
  • BEH technology ensures high pH dissolution resistance and enhanced chemical stability.
  • Reduced USP peak tailing factors for bases.
  • Improved low-pH stability and virtually eliminates LC/MS bleed.
Catalog No. 50-781-612