Learn More
Waters Corp Waters XBridge BEH C8 Column, 130Å, 3.5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186003055
Encompass_Preferred
The XBridge BEH C8 column features a trifunctionally bonded C8 sorbent, less retentive than C18, for faster analysis. It offers a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed, enabling robust HPLC separations with improved peak shape.
  • Offers flexibility for rapid, robust HPLC methods development.
  • Works over a wide pH range (1 to 12) and unprecedented temperature ranges.
  • Provides seamless method migration to UPLC.
  • Ensures unmatched column lifetime via trifunctional bonding and advanced end-capping.
  • Features improved low pH stability and chemical stability at high pH.
  • Allows switching between high and low pH on a single column.
  • Consistent manufacturing ensures batch reproducibility.
  • Delivers improved performance and long column lifetime under demanding LC conditions.
Catalog No. 50-781-508