Learn More
Waters Corp Column, XBridge Glycan BEH Amide, 2.5 μm, 3 mm, 75 mm
Supplier:  Waters Corp 186008039
Encompass_Preferred
The XBridge Glycan BEH Amide Column is designed for hydrophilic interaction (HILIC) separations, ideal for highly polar substances like carbohydrates and sugars. It features a stable, trifunctionally bonded amide phase and ethylene bridged hybrid (BEH) particle technology for adaptability across a wide pH range.
  • Available in 1.7μm, 2.5μm XP, and 3.5μm particle sizes for UPLC and HPLC.
  • Batch-to-batch consistency ensured by quality control testing with the 2-AB labeled Glycan Performance standard.
  • Chemically stable, trifunctionally bonded amide phase maintains stability across a wide pH range.
  • BEH particle technology provides exceptional stability and adaptability in HILIC separations.
  • Shares selectivity with ACQUITY UPLC BEH Amide columns for easy separation transfer between HPLC and UPLC.
  • Effectively separates very polar substances, including carbohydrates and sugars.
Catalog No. 50-239-2627