Learn More
Waters Corp HILIC Column, 130 Å, 5 μm, 3 mm x 50 mm
Supplier:  Waters Corp 186004447
Encompass_Preferred
Columns incorporate unbonded BEH substrate, designed for the retention and separation of very polar compounds. These columns are optimized and tested to deliver efficient and reproducible separations under HPLC HILIC conditions. They are more rugged than silica-based HILIC phases due to the chemical stability of the BEH particle, resulting in extended column lifetimes.
  • Contain unbonded BEH substrate and are designed to retain and separate very polar compounds.
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica based HILIC phases; the BEH particle’s chemical stability results in long column lifetimes.
Catalog No. 50-782-426