Learn More
Waters Corp XBridge Peptide BEH C18 OBD Prep Column, 10 μm
Supplier: Waters Corp 186008194
XBridge Peptide BEH C18 OBD Prep Columns are designed for efficient peptide separations. Featuring a small-pore (130Å) trifunctionally bonded BEH particle, these columns provide a wide pH range, excellent low pH stability, and minimal column bleed. They are ideal for proteomics, protein characterization, and peptide synthesis applications.
- Optimized and QC tested for peptide separations
- Small-pore (130Å) trifunctionally bonded BEH particle
- Widest usable pH range (1 to 12)
- Superior low pH stability
- Ultra-low column bleed
- C18 chemistry and hybrid particle substrate
- 10 μm particle size and 130 Å pore size
- Dimensions: 10 mm inner diameter × 50 mm length
- Maximum pressure: 6000 psi (415 Bar)
- Compatible with HPLC systems
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.