Learn More
Waters Corp Prep Column, C18, 130Å, 10 μm, 10 mm × 250 mm
Supplier: Waters Corp 186008197
XBridge Peptide BEH C18 columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
- C18 chemistry
- Reversed phase separation mode
- Hybrid particle substrate
- pH range from 1 to 12
- Maximum pressure of 6000 psi (415 Bar)
- Endcapped
- Low silanol activity
- Spherical particle shape
- 10 μm particle size
- 130 Å pore size
- Peptide QC tested
- OBD prep column format
- HPLC system compatibility
- BEH particle technology
- L1 USP classification
- 10 mm inner diameter
- 250 mm length
- 18% carbon load
- Designed for peptide applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.