Learn More
Waters Corp Waters Corporation Preparative Column, 300Å, 10 μm, 10 mm × 100 mm
Supplier: Waters Corp 186008199
These Peptide BEH C18 columns are optimized and QC tested for peptide separations, offering a wide usable pH range and superior stability for proteomics and peptide synthesis applications.
- Optimized and QC tested for peptide separations, featuring wide-pore (300Å) BEH particles.
- Offers the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
- Enables direct scaling from UPLC or HPLC screening to lab-scale purification.
- Provides good peak shape, high efficiency, and outstanding stability.
- Chemistry: C18; Separation mode: Reversed Phase; Particle substrate: Hybrid.
- Particle size: 10 μm; Pore size: 300 Å.
- Dimensions: 10 mm inner diameter, 100 mm length.
- Maximum pressure: 6000 psi (415 Bar).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.