Learn More
Waters Corp XBridge Peptide BEH C18 Prep Guard Cartridge, 300 Å, 10 μm, 30 mm x 10 mm
Supplier:  Waters Corp 186006882
Encompass_Preferred
This Prep Guard Cartridge is designed to protect both HPLC and SFC 30 mm ID Prep columns. Placing this guard assembly before the prep column extends column lifetimes and reduces purification costs.
  • Protects HPLC and SFC 30 mm ID prep columns when used with a guard holder.
  • Recommended for use between the injector and main column with matching chemistry and particle size.
  • Packing materials provide excellent peak shape, high efficiency, and stability for acidic and basic mobile phases.
  • Produced in a cGMP, ISO 9001:2000 certified facility using ultra-pure reagents.
  • Each batch is chromatographically tested with acidic, basic, and neutral analytes for repeatable performance.
  • Manufacturing process is meticulously monitored and controlled to ensure high quality.
Catalog No. 50-239-1369