Learn More
Waters Corp Column, XBridge Peptide BEH C18 XP, 300Å Pore, 2.5μm Particle, 4.6 mm x 150 mm
Supplier:  Waters Corp 186009076
Encompass_Preferred
The XBridge Peptide BEH C18 XP Column is optimized and QC tested for peptide separations. It features a 300Å pore size trifunctionally bonded BEH particle, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed for large peptides in various applications.
  • Optimized and QC tested for peptide separations.
  • Features 300Å pore size trifunctionally bonded BEH particle.
  • Offers the widest usable pH range.
  • Provides superior low pH stability.
  • Ensures ultra-low column bleed for especially large peptides.
  • Suitable for proteomic, peptide mapping, and synthetic peptide applications.
  • Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c.
Catalog No. 50-239-1232