Learn More
Waters Corp XBridge BEH Phenyl Column, 130Å, 5 µm, 3 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003345
Encompass_Preferred
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Catalog No. 50-781-697