Learn More
Waters Corp XBridge Premier BEH Amide Column, 2.5 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186009928
Encompass_Preferred
XBridge Premier Columns are designed for ultra-high-performance liquid chromatography (UHPLC), integrating Ethylene Bridged Hybrid (BEH) particles with MaxPeak High Performance Surfaces (HPS) Technology for enhanced chromatographic performance and stability.
  • Improved analyte sensitivity and peak shapes.
  • High pH stability (1 to 12) and chemical resistance.
  • MaxPeak HPS Technology minimizes analyte/surface interactions, improving recovery and purity.
  • BEH Technology provides better peak shape for bases and a wider operating pH range.
  • Robust mechanical strength for efficient UHPLC operation.
  • Optional VanGuard FIT for extended column life and easy installation.
  • eCord/eConnect options for column tracking and compliance.
Catalog No. NC3875387