Learn More
Waters Corp XBridge Premier BEH C18 Column, 3.5 μm, 2.1 x 100 mm
Supplier: Waters Corp 186010651
This column with a 3.5 μm particle size and 2.1 x 100 mm dimensions, offers a trifunctionally bonded C18 ligand for universal LC separations. It provides a wide usable pH range of 1-12, superior low pH stability, and ultra-low column bleed. MaxPeak High Performance Surface hardware technology minimizes unwanted analyte/surface interactions, improving peak shape and signal intensity for HPLC assays and UPLC transitions.
- C18 chemistry, reversed phase separation
- Hybrid spherical particle substrate
- pH range of 1 to 10
- Temperature limits: 80 °C (low pH) to 60 °C (high pH)
- Maximum pressure: 6000 psi (415 Bar)
- Particle size: 3.5 μm; pore size: 130 Å
- MaxPeak High Performance Surface reduces analyte/surface interactions
- BEH technology for superior peak shape and wide pH stability
- Improved sensitivity, recovery, and reduced conditioning times
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.