Learn More
Waters Corp Waters XBridge Column, C8, 2.5 μm, 4.6 mm, 50 mm
Supplier:  Waters Corp 186010370
Encompass_Preferred
These columns are less retentive than C18 columns, ideal for fast method development. They feature trifunctionally bonded BEH particles, offering a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss.
  • Less retentive than C18 columns, aiding fast method development.
  • Trifunctionally bonded BEH particles provide a wide usable pH range (1 to 12).
  • Offers superior low pH stability.
  • Ensures ultra-low column bleed.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions.
  • Helps prevent poor peak shape and signal intensity losses.
Catalog No. 50-226-9378