Learn More
Waters Corp XBridge Premier BEH Phenyl Column, 3.5 μm, 4.6 x 100 mm, 1 pk
Supplier:  Waters Corp 186010676
Encompass_Preferred
These XBridge Premier BEH Phenyl 3.5 μm, 4.6 x 100 mm columns offer complementary selectivity for polyaromatic compounds. Featuring a highly stable phenyl sorbent, they ensure ultra-low column bleed, extended lifetimes, and excellent peak shape. MaxPeak Premier hardware minimizes analyte/surface interactions, preventing poor peak shape and signal loss.
  • Reversed phase separation.
  • Hybrid, spherical particles with low silanol activity.
  • pH range of 1 to 12; temperature limits up to 80°C (low pH) or 60°C (high pH).
  • Maximum pressure: 6000 psi (415 bar).
  • Endcapped with phenyl-hexyl bonding technology.
  • 130 Å pore size and 185 surface area.
  • Parker-style endfitting.
  • Designed for HPLC and LC, USP classification L11.
  • 15% carbon load, suitable for small molecules.
Catalog No. 50-288-6235