Learn More
Waters Corp XBridge Premier BEH Phenyl Column, 3.5 μm, 4.6 mm, 150 mm, 1 pk
Supplier:  Waters Corp 186010677
Encompass_Preferred
These columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. The column's sorbent is the most chemically stable phenyl sorbent available, providing ultra-low column bleed, extended column lifetimes, and excellent peak shape. MaxPeak High Performance Surface hardware technology is incorporated to significantly reduce unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.
  • Provides complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds.
  • Features a chemically stable phenyl sorbent, offering ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Utilizes MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions, which can cause poor peak shape and signal intensity loss.
Catalog No. 50-288-6248