Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Premier BEH Phenyl VanGuard FIT Column, 2.5 μm, 4.6 x 100 mm

Supplier:  Waters Corp 186010353

Encompass_Preferred

This column is designed for high-performance liquid chromatography applications. It features advanced BEH Technology and Phenyl chemistry for robust and reliable separations, incorporating MaxPeak High Performance Surfaces.

  • Features BEH Technology
  • Utilizes Phenyl chemistry
  • Incorporates MaxPeak High Performance Surfaces
  • Includes VanGuard FIT integrated guard column technology that maintains separation efficiency
  • Extends column lifetime
  • Particle size: 2.5 μm
  • Dimensions: 4.6 x 100 mm
  • Pore diameter: 130 Å

Catalog No. 50-226-9361


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.