Learn More
Waters Corp Waters Column, XBridge Premier BEH Shield RP18, 2.5 μm, 4.6 mm X 50 mm
Supplier:  Waters Corp 186009921
Encompass_Preferred
XBridge Premier BEH Shield RP18 Columns complement Premier phases for method development, offering alternate selectivity for phenolic compounds via an embedded carbamate group. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity losses.
  • Minimizes analyte/surface interactions, improving recovery, sensitivity, and reproducibility.
  • Mitigates loss of metal-sensitive analytes (lipids, organic acids, peptides, oligonucleotides).
  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes.
  • Simplifies mobile phases by reducing need for complex additives.
  • Saves time in method development and increases data confidence.
Catalog No. NC3985576