Learn More
Waters Corp XBridge Premier Oligonucleotide BEH C18 Column, 300 Å, 2.5 μm, 4.6 mm X 100 mm
Supplier:  Waters Corp 186010546
Encompass_Preferred
This column is designed for the separation of oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UHPLC/HPLC systems. It incorporates BEH hybrid particle technology for pH and temperature stability and MaxPeak Premier high performance surface technology for ultra-low non-specific adsorption. Stationary phase batches are QC tested to ensure lot-to-lot performance, offering sensitivity, reproducibility, and excellent out-of-the-box performance.
  • BEH hybrid particle technology provides unmatched pH and temperature stability.
  • MaxPeak Premier high performance surface technology offers ultra-low non-specific adsorption.
  • QC tested stationary phase batches ensure lot-to-lot performance.
  • Delivers unmatched sensitivity, exquisite reproducibility, and excellent out-of-the-box performance.
  • 300Å pore size columns offer enhanced resolution for longer oligonucleotides.
Catalog No. 50-239-1196