Learn More
Waters Corp Peptide Column, C18, 130 Å, 2.5 μm
Supplier:  Waters Corp 186010703
Encompass_Preferred
This XBridge Premier Peptide BEH C18 Column is optimized for peptide separations, featuring 130 Å pore size, 2.5 μm particle size, and 4.6 mm × 50 mm dimensions. It utilizes MaxPeak High Performance Surfaces technology and includes a Premier VanGuard FIT pre-column for extended life. Compatible with UHPLC and UPLC systems.
  • Optimized and QC tested for peptide separations with cytochrome c digest standards.
  • BEH particle offers wide pH range, low pH stability, and ultra-low column bleed.
  • MaxPeak Premier class and HPS technology enhance reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Integrated 5mm pre-column protects and extends analytical column life.
  • Reduces conditioning, simplifies mobile phases, and improves sensitivity.
Catalog No. 50-239-1124