Learn More
Waters Corp Column, Peptide BEH C18, 130 Å, 2.5 μm, 4.6 x 150 mm
Supplier:  Waters Corp 186010705
Encompass_Preferred
The XBridge Premier Peptide BEH C18 Column is engineered for high-performance peptide separations. It features MaxPeak High Performance Surfaces (HPS) technology, minimizing analyte/surface interactions to improve reproducibility, peak shape, and recovery for metal-sensitive analytes in proteomics, protein characterization, and peptide synthesis.
  • Optimized for peptide separations, offering wide pH range and low column bleed.
  • MaxPeak HPS technology enhances analyte recovery, sensitivity, and reproducibility.
  • Integrated 5 mm pre-column extends analytical column life without compromising resolution.
  • BEH Technology™ ensures excellent peak shape and stability across broad pH and temperature ranges.
Catalog No. 50-239-2568