Learn More
Waters Corp XBridge Premier Protein BEH C4 Column, 300 Å, 2.5 μm, 2.1 x 50 mm
Supplier:  Waters Corp 176005110
Encompass_Preferred
The XBridge Premier Protein BEH C4 column, with a 300 Å pore size and 2.5 μm particle size, is designed to enhance protein analysis by minimizing analyte/surface interactions, thereby increasing analyte recovery, sensitivity, and reproducibility. This column features proprietary monofunctional C4 bonding to an Ethylene Bridged Hybrid (BEH) substrate and utilizes MaxPeak High Performance Surfaces.
  • Minimizes analyte/surface interactions, increasing analyte recovery, sensitivity, and reproducibility.
  • Separates proteins of various sizes, hydrophobicities, and isoelectric points.
  • Tolerates extreme pH and temperature.
  • Minimal secondary interactions.
  • QC batch-tested with the MassPREP Protein Mix Standard.
  • Utilizes MaxPeak high performance surfaces.
  • Hybrid inorganic/organic LC surfaces to protect metal-sensitive analytes.
Catalog No. 50-239-1615