Learn More
Waters Corp Column, XBridge Premier Protein BEH C4, 300 Å, 2.5 μm
Supplier:  Waters Corp 176005114
Encompass_Preferred
MaxPeak Premier Columns enhance chromatographic separations by reducing the loss of metal-sensitive analytes. Featuring MaxPeak High Performance Surfaces (HPS) technology, these columns minimize analyte/surface interactions for improved results. The XBridge Premier Protein BEH C4, 300 Å column is engineered for protein analysis, offering separation based on size, hydrophobicity, and isoelectric points, with tolerance to extreme pH and temperature conditions.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk and greater confidence in data and decision making.
Catalog No. 50-239-2891