Learn More
Waters Corp SEC Column, 250 Å, 2.5 μm, 7.8 x 150 mm
Supplier:  Waters Corp 186009961
Encompass_Preferred
XBridge Premier Protein SEC Columns assist scientists in achieving reliable protein size variant separations, covering a range of 10,000 to approximately 650,000 Daltons. This is accomplished through the combined use of MaxPeak Premier High Performance Surfaces and innovative BEH-PEO Particle technologies.
  • Minimize unwanted secondary ionic or hydrophobic interactions between proteins and the column.
  • Reduce need for extensive method development.
  • Enable utilization of a "generic" or "platform-type" method for a wide array of samples.
  • Each batch is specifically QC tested using mAb Size Variant Standard to ensure consistent performance.
  • Designed for UPLC SEC applications with a compatible low LC system volume.
  • Suitable for gel filtration, size exclusion chromatography, aggregate, monomer and fragment analysis, and antibody conjugates.
Catalog No. 50-239-2710