Learn More
Waters Corp XBridge Premier Protein SEC Column, 250 Å, 2.5 μm, 7.8 x 300 mm
Supplier:  Waters Corp 186009962
Encompass_Preferred
XBridge Premier Protein SEC Columns offer reliable protein size variant separations (10,000 to approximately 650,000 Daltons). Featuring MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies, these columns minimize non-desired ionic or hydrophobic interactions, simplifying method development. Each batch is QC tested for consistent performance in UPLC SEC applications, requiring a low LC system volume.
  • Separates protein size variants (10,000 to 650,000 Daltons)
  • Minimizes ionic/hydrophobic interactions
  • Simplifies method development (generic/platform methods)
  • QC tested for consistent performance
  • Designed for UPLC SEC with low LC system volume
  • Applications: gel filtration, size exclusion, aggregate, monomer, fragment analysis, antibody conjugates
Catalog No. 50-239-0531