Learn More
Waters Corp SEC Column, 250 Å, 2.5 μm, 7.8 mm ID, 300 mm Length, with MaxPeak Premier Protein SEC Guard
Supplier:  Waters Corp 176004793
Encompass_Preferred
This SEC column provides reliable protein size variant separations by combining MaxPeak Premier High Performance Surfaces and novel BEH-PEO Particle technologies. It minimizes undesired secondary interactions, which reduces method development time and improves data confidence for complex biotherapeutics.
  • Achieves reliable protein size variant separations from 10,000 to approximately 650,000 Daltons.
  • Minimizes undesired secondary ionic or hydrophobic interactions.
  • Reduces method development by enabling a “generic” approach for various samples.
  • Increases analyte recovery, sensitivity, and reproducibility with MaxPeak High Performance Surfaces technology.
  • Reduces column conditioning and passivation times.
  • Allows for simpler mobile phases without complex additives.
  • Lowers the cost per analysis by utilizing Premier SEC Guards.
Catalog No. 50-239-0691