Learn More
Waters Corp XBridge Premier Protein SEC Column, MaxPeak Premier Protein SEC Guard, 250Å, 2.5 μm
Supplier:  Waters Corp 176004782
Encompass_Preferred
XBridge Premier Protein SEC 250Å, 2.5 μm Columns provide reliable protein size variant separations (10,000 to approximately 650,000 Daltons). Waters MaxPeak Premier High Performance Surfaces and novel BEH-PEO Particle technologies work synergistically to minimize unwanted secondary ionic or hydrophobic interactions, reducing costly method development by enabling “generic” or “platform-type” methods for diverse samples.
  • Reliable protein size variant separations for proteins from 10,000 to approximately 650,000 Daltons.
  • Minimizes non-desired interactions using MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies.
  • Reduces method development time with “generic” or “platform-type” methods.
  • Includes a MaxPeak Premier Protein SEC Guard (250Å, 2.5 μm, 4.6 x 30 mm).
Catalog No. 50-239-1658